Anti-DLAT Mouse Monoclonal Antibody
BWRLMB0031
- Antibody type:Primary
- Antigen symbol:DLAT
- Clonality:Monoclonal
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human DLAT.
This antibody detects endogenous levels of Pyruvate Dehydrogenase E2 and does not cross-react with related proteins
Type: Primary
Antigen: DLAT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse