Anti-ALDH3A2 Mouse Monoclonal Antibody
Catalog # BWRLMB0022
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ALDH3A2
- Clonality:Monoclonal
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human ALDH3A2.
This antibody detects endogenous levels of ALDH3A2 and does not cross-react with related proteins
Type: Primary
Antigen: ALDH3A2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human