Anti-TADA3L Rabbit Polyclonal Antibody
BWRLBS7973
- Antibody type:Primary
- Antigen symbol:TADA3L
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human TADA3L.
TADA3L pAb detects endogenous levels of TADA3L protein.
Type: Primary
Antigen: TADA3L
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat