Order Entry
Italy
ContactUsLinkComponent
Anti-Nkx-2.8 Rabbit Polyclonal Antibody [clone: P37]
Anti-Nkx-2.8 Rabbit Polyclonal Antibody [clone: P37]
Catalog # BWRLBS9251
Anti-Nkx-2.8 Rabbit Polyclonal Antibody [clone: P37]
Catalog # BWRLBS9251

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Nkx-2.8
  • Clonality:
    Polyclonal
  • Clone:
    P37
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 11-60 of Human Nkx-2.8.

Nkx-2.8 (P37) pAb detects endogenous levels of Nkx-2.8 protein.

Type: Primary
Antigen: Nkx-2.8
Clonality: Polyclonal
Clone: P37
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human