Anti-Microcephalin Rabbit Polyclonal Antibody [clone: F131]
BWRLBS9224
- Antibody type:Primary
- Antigen symbol:Microcephalin
- Clonality:Polyclonal
- Clone:F131
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 100-150 of Human Microcephalin.
Microcephalin (F131) pAb detects endogenous levels of Microcephalin protein.
Type: Primary
Antigen: Microcephalin
Clonality: Polyclonal
Clone: F131
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse