Anti-STK33 Rabbit Polyclonal Antibody [clone: N108]
Catalog # BWRLBS9197
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:STK33
- Clonality:Polyclonal
- Clone:N108
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 72-124 of Human STK33.
STK33 (N108) pAb detects endogenous levels of STK33 protein.
Type: Primary
Antigen: STK33
Clonality: Polyclonal
Clone: N108
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human