Order Entry
Italy
ContactUsLinkComponent
Anti-MMP-10 Rabbit Polyclonal Antibody
Anti-MMP-10 Rabbit Polyclonal Antibody
Catalog # BWRLBS7167
Anti-MMP-10 Rabbit Polyclonal Antibody
Catalog # BWRLBS7167

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MMP-10
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recombinant full length Human MMP-10.

MMP-10 pAb detects endogenous levels of MMP-10 protein.

Type: Primary
Antigen: MMP-10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human