Anti-p-Myc Rabbit Polyclonal Antibody [clone: S62]
Catalog # BWRLBS4126
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Myc
- Clonality:Polyclonal
- Clone:S62
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Myc around the phosphorylation site of Serine 62.
p-Myc (S62) pAb detects endogenous levels of proto-oncogene c-Myc protein only when phosphorylated at Ser62.
Type: Primary
Antigen: p-Myc
Clonality: Polyclonal
Clone: S62
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat