Anti-CHST1 Rabbit Polyclonal Antibody
BWRLBS5662
- Antibody type:Primary
- Antigen symbol:CHST1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 126-171 of Human CHST1.
CHST1 pAb detects endogenous levels of CHST1 protein.
Type: Primary
Antigen: CHST1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat