Order Entry
Italy
ContactUsLinkComponent
Anti-p-IkB-beta Rabbit Polyclonal Antibody [clone: S23]
Anti-p-IkB-beta Rabbit Polyclonal Antibody [clone: S23]
Catalog # BWRLBS4106
Anti-p-IkB-beta Rabbit Polyclonal Antibody [clone: S23]
Catalog # BWRLBS4106

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-IkB- beta
  • Clonality:
    Polyclonal
  • Clone:
    S23
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human IkB-beta around the phosphorylation site of Serine 23.

p-I?B-? (S23) pAb detects endogenous levels of I?B-? protein only when phosphorylated at Ser23.

Type: Primary
Antigen: p-IkB-beta
Clonality: Polyclonal
Clone: S23
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat