Anti-Stat5b Rabbit Polyclonal Antibody
Catalog # BWRLBS6031
Nome del fornitore: Bioworld Technology
Specifiche
- Antibody type:Primary
- Antigen symbol:Stat5b
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifiche
Informazioni su questo articolo
Recombinant full length Human Stat5b.
STAT5B pAb detects endogenous levels of STAT5B protein.
Type: Primary
Antigen: Stat5b
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat