Anti-p-IGF-1R Rabbit Polyclonal Antibody [clone: Y980]
Catalog # BWRLBS5081
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-IGF-1R
- Clonality:Polyclonal
- Clone:Y980
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human IGF-1R around the phosphorylation site of Tyrosine 980.
p-IGF1R (Y980) pAb detects endogenous levels of IGF1R protein only when phosphorylated at Tyr980.
Type: Primary
Antigen: p-IGF-1R
Clonality: Polyclonal
Clone: Y980
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat