Anti-p-HDAC1 Rabbit Polyclonal Antibody [clone: S421/423]
BWRLBS5077
- Antibody type:Primary
- Antigen symbol:p-HDAC1
- Clonality:Polyclonal
- Clone:S421/423
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human HDAC1 around the phosphorylation site of Serine 421/423.
p-HDAC1 (S421/423) pAb detects endogenous levels of HDAC1 protein only when phosphorylated at Ser421/423.
Type: Primary
Antigen: p-HDAC1
Clonality: Polyclonal
Clone: S421/423
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat