Anti-p-Synapsin I Rabbit Polyclonal Antibody [clone: S62]
Catalog # BWRLBS4879
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Synapsin I
- Clonality:Polyclonal
- Clone:S62
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Synapsin I around the phosphorylation site of Serine 62.
p-Synapsin I (S62) pAb detects endogenous levels of Synapsin I protein when phosphorylated at Ser62.
Type: Primary
Antigen: p-Synapsin I
Clonality: Polyclonal
Clone: S62
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat