Anti-p-SLP-76 Rabbit Polyclonal Antibody [clone: Y128]
Catalog # BWRLBS4872
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-SLP-76
- Clonality:Polyclonal
- Clone:Y128
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human SLP-76 around the phosphorylation site of Tyrosine 128.
p-SLP-76 (Y128) pAb detects endogenous levels of SLP-76 protein only when phosphorylated at Tyr128.
Type: Primary
Antigen: p-SLP-76
Clonality: Polyclonal
Clone: Y128
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat