Anti-Ataxin-10 Rabbit Polyclonal Antibody
BWRLBS60104
- Antibody type:Primary
- Antigen symbol:Ataxin-10
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
A synthetic peptide corresponding to residues in Human Ataxin-10.
Ataxin-10 pAb detects endogenous levels of Ataxin-10 protein.
Type: Primary
Antigen: Ataxin-10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat