Anti-Ghrelin Rabbit Polyclonal Antibody
BWRLBS6800
- Antibody type:Primary
- Antigen symbol:Ghrelin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Recombinant full length Human Ghrelin.
Ghrelin pAb detects endogenous levels of Ghrelin protein.
Type: Primary
Antigen: Ghrelin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat