Order Entry
Italy
ContactUsLinkComponent
Anti-p-DARPP-32 Rabbit Polyclonal Antibody [clone: T34]
Anti-p-DARPP-32 Rabbit Polyclonal Antibody [clone: T34]
Catalog # BWRLBS4782
Anti-p-DARPP-32 Rabbit Polyclonal Antibody [clone: T34]
Catalog # BWRLBS4782

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-DARPP-32
  • Clonality:
    Polyclonal
  • Clone:
    T34
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human DARPP-32 around the phosphorylation site of Threonine 34.

p-DARPP-32 (T34) pAb detects endogenous levels of DARPP-32 protein only when phosphorylated at Thr34.

Type: Primary
Antigen: p-DARPP-32
Clonality: Polyclonal
Clone: T34
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat