Anti-p-CPI-17 Rabbit Polyclonal Antibody [clone: T38]
BWRLBS4780
- Antibody type:Primary
- Antigen symbol:p-CPI-17
- Clonality:Polyclonal
- Clone:T38
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human CPI-17 around the phosphorylation site of Threonine 38.
p-CPI-17 (T38) pAb detects endogenous levels of CPI-17 pAb protein only when phosphorylated at Thr38.
Type: Primary
Antigen: p-CPI-17
Clonality: Polyclonal
Clone: T38
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat