Anti-POLR1C Rabbit Polyclonal Antibody
Catalog # BWRLBS5916
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:POLR1C
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 146-191 of Human POLR1C.
POLR1C pAb detects endogenous levels of POLR1C protein.
Type: Primary
Antigen: POLR1C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat