Order Entry
Italy
ContactUsLinkComponent
Anti-p-Bcl-2 Rabbit Polyclonal Antibody [clone: T69]
Anti-p-Bcl-2 Rabbit Polyclonal Antibody [clone: T69]
Catalog # BWRLBS4294
Anti-p-Bcl-2 Rabbit Polyclonal Antibody [clone: T69]
Catalog # BWRLBS4294

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-Bcl-2
  • Clonality:
    Polyclonal
  • Clone:
    T69
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human Bcl-2 around the phosphorylation site of Threonine 69.

P-Bcl-2 (T69) pAb detects endogenous levels of Bcl-2 protein only when phosphorylated at Thr69 . .

Type: Primary
Antigen: p-Bcl-2
Clonality: Polyclonal
Clone: T69
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat