Anti-p16 INK4a Rabbit Polyclonal Antibody
BWRLBS6431
- Antibody type:Primary
- Antigen symbol:p16 INK4a
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Recombinant full length Human p16 INK4a.
p16 INK4a pAb detects endogenous levels of p16 INK4a protein.
Type: Primary
Antigen: p16 INK4a
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human