Order Entry
Italy
ContactUsLinkComponent
Anti-MRP-S27 Rabbit Polyclonal Antibody [clone: Y48]
Anti-MRP-S27 Rabbit Polyclonal Antibody [clone: Y48]
Catalog # BWRLBS3326
Anti-MRP-S27 Rabbit Polyclonal Antibody [clone: Y48]
Catalog # BWRLBS3326

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MRP-S27
  • Clonality:
    Polyclonal
  • Clone:
    Y48
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 16-62 of Human MRP-S27.

MRP-S27 (Y48) pAb detects endogenous levels of MRP-S27 protein.

Type: Primary
Antigen: MRP-S27
Clonality: Polyclonal
Clone: Y48
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse