Order Entry
Italy
ContactUsLinkComponent
Anti-Ku-70 Rabbit Polyclonal Antibody [clone: S1]
Anti-Ku-70 Rabbit Polyclonal Antibody [clone: S1]
Catalog # BWRLBS1818
Anti-Ku-70 Rabbit Polyclonal Antibody [clone: S1]
Catalog # BWRLBS1818

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Ku-70
  • Clonality:
    Polyclonal
  • Clone:
    S1
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to the N-terminus of Human Ku-70.

Ku70 (S1) pAb detects endogenous levels of Ku70 protein.

Type: Primary
Antigen: Ku-70
Clonality: Polyclonal
Clone: S1
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse