Anti-MRG15 Rabbit Polyclonal Antibody [clone: R56]
BWRLBS2674
- Antibody type:Primary
- Antigen symbol:MRG15
- Clonality:Polyclonal
- Clone:R56
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 23-75 of Human MRG15.
MRG15 (R56) pAb detects endogenous levels of MRG15 protein.
Type: Primary
Antigen: MRG15
Clonality: Polyclonal
Clone: R56
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse