Anti-PKAalpha/beta cat Rabbit Polyclonal Antibody [clone: K17]
Catalog # BWRLBS2648
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PKA alpha/beta
- Clonality:Polyclonal
- Clone:K17
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminus of Human PKAalpha.
PKA?/? cat (K17) pAb detects endogenous levels of PKA?/? protein.
Type: Primary
Antigen: PKAalpha/beta cat
Clonality: Polyclonal
Clone: K17
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat