Anti-RPL12 Rabbit Polyclonal Antibody [clone: N103]
BWRLBS3046
- Antibody type:Primary
- Antigen symbol:RPL12
- Clonality:Polyclonal
- Clone:N103
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 72-121of Human RPL12.
RPL12 (N103) pAb detects endogenous levels of Ribosomal Protein L12 protein.
Type: Primary
Antigen: RPL12
Clonality: Polyclonal
Clone: N103
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat