Anti-CYP7B1 Rabbit Polyclonal Antibody [clone: Q127]
Catalog # BWRLBS3024
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CYP7B1
- Clonality:Polyclonal
- Clone:Q127
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 101-150 of Human CYP7B1.
CYP7B1 (Q127) pAb detects endogenous levels of CYP7B1 protein.
Type: Primary
Antigen: CYP7B1
Clonality: Polyclonal
Clone: Q127
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat