Order Entry
Italy
ContactUsLinkComponent
Anti-Cox-17 Rabbit Polyclonal Antibody [clone: N8]
Anti-Cox-17 Rabbit Polyclonal Antibody [clone: N8]
Catalog # BWRLBS2514
Anti-Cox-17 Rabbit Polyclonal Antibody [clone: N8]
Catalog # BWRLBS2514

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Cox-17
  • Clonality:
    Polyclonal
  • Clone:
    N8
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to the N-terminal of Human Cox-17.

COX17 (N8) pAb detects endogenous levels of COX17 protein.

Type: Primary
Antigen: Cox-17
Clonality: Polyclonal
Clone: N8
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat