Order Entry
Italy
ContactUsLinkComponent
Anti-XIAP Rabbit Polyclonal Antibody [clone: G81]
Anti-XIAP Rabbit Polyclonal Antibody [clone: G81]
Catalog # BWRLBS1609
Anti-XIAP Rabbit Polyclonal Antibody [clone: G81]
Catalog # BWRLBS1609

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    XIAP
  • Clonality:
    Polyclonal
  • Clone:
    G81
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 50-100 of Human XIAP.

XIAP (G81) pAb detects endogenous levels of XIAP protein.

Type: Primary
Antigen: XIAP
Clonality: Polyclonal
Clone: G81
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat