Anti-Midline-1 Rabbit Polyclonal Antibody [clone: E103]
Catalog # BWRLBS2451
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Midline-1
- Clonality:Polyclonal
- Clone:E103
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 71-120 of Human Midline-1.
Midline-1 (E103) pAb detects endogenous levels of Midline-1 protein.
Type: Primary
Antigen: Midline-1
Clonality: Polyclonal
Clone: E103
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat