Anti-C1q-C Rabbit Polyclonal Antibody [clone: Q119]
Catalog # BWRLBS3838
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:C1q-C
- Clonality:Polyclonal
- Clone:Q119
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 81-130 of Human C1q-C.
C1q-C (Q119) pAb detects endogenous levels of C1q-C protein.
Type: Primary
Antigen: C1q-C
Clonality: Polyclonal
Clone: Q119
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat