Order Entry
Italy
ContactUsLinkComponent
Anti-SLP-76 Rabbit Polyclonal Antibody [clone: G122]
Anti-SLP-76 Rabbit Polyclonal Antibody [clone: G122]
Catalog # BWRLBS3758
Anti-SLP-76 Rabbit Polyclonal Antibody [clone: G122]
Catalog # BWRLBS3758

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SH2 Domain Containing leukocyte Protein of 76kDa
  • Clonality:
    Polyclonal
  • Clone:
    G122
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 100-150 of Human SLP-76.

SLP-76 (G122) pAb detects endogenous levels of SLP-76 protein.

Type: Primary
Antigen: SLP-76
Clonality: Polyclonal
Clone: G122
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat