Anti-Translin Rabbit Polyclonal Antibody [clone: E121]
Catalog # BWRLBS2314
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Translin
- Clonality:Polyclonal
- Clone:E121
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 88-146 of Human Translin.
Translin (E121) pAb detects endogenous levels of Translin protein.
Type: Primary
Antigen: Translin
Clonality: Polyclonal
Clone: E121
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat