Anti-SENP3 Rabbit Polyclonal Antibody [clone: Y24]
BWRLBS1471
- Antibody type:Primary
- Antigen name:Sentrin-specific protease 3
- Antigen symbol:SENP3
- Clonality:Polyclonal
- Clone:Y24
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminual of Human SENP3.
SENP3 (Y24) pAb detects endogenous levels of SENP3 protein.
Type: Primary
Antigen: SENP3
Clonality: Polyclonal
Clone: Y24
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse