Anti-MEK-7 Rabbit Polyclonal Antibody [clone: I264]
BWRLBS3747
- Antibody type:Primary
- Antigen symbol:MEK-7
- Clonality:Polyclonal
- Clone:I264
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 241-290 of Human MEK-7.
MEK-7 (I264) pAb detects endogenous levels of MEK-7 protein.
Type: Primary
Antigen: MEK-7
Clonality: Polyclonal
Clone: I264
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat