Anti-Prefoldin 1 Rabbit Polyclonal Antibody [clone: E35]
Catalog # BWRLBS2245
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Prefoldin 1
- Clonality:Polyclonal
- Clone:E35
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 11-60 of Human Prefoldin 1.
Prefoldin 1 (E35) pAb detects endogenous levels of Prefoldin 1 protein.
Type: Primary
Antigen: Prefoldin 1
Clonality: Polyclonal
Clone: E35
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse