Order Entry
Italy
ContactUsLinkComponent
Anti-Cadherin-16 Rabbit Polyclonal Antibody [clone: M810]
Anti-Cadherin-16 Rabbit Polyclonal Antibody [clone: M810]
Catalog # BWRLBS2221
Anti-Cadherin-16 Rabbit Polyclonal Antibody [clone: M810]
Catalog # BWRLBS2221

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Cadherin-16
  • Clonality:
    Polyclonal
  • Clone:
    M810
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 791-840 of Human Cadherin-16.

Cadherin-16 (M810) pAb detects endogenous levels of Cadherin-16 protein.

Type: Primary
Antigen: Cadherin-16
Clonality: Polyclonal
Clone: M810
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat