Order Entry
Italy
ContactUsLinkComponent
Anti-CYP2C8/9/18/19 Rabbit Polyclonal Antibody [clone: M136]
Anti-CYP2C8/9/18/19 Rabbit Polyclonal Antibody [clone: M136]
Catalog # BWRLBS2195
Anti-CYP2C8/9/18/19 Rabbit Polyclonal Antibody [clone: M136]
Catalog # BWRLBS2195

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CYP2C8/9/18/19
  • Clonality:
    Polyclonal
  • Clone:
    M136
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 100-150 of Human CYP2C8.

CYP2C8/9/18/19 (M136) pAb detects endogenous levels of CYP2C8/9/18/19 protein.

Type: Primary
Antigen: CYP2C8/9/18/19
Clonality: Polyclonal
Clone: M136
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat