Anti-CstF-50 Rabbit Polyclonal Antibody [clone: R3]
BWRLBS2036
- Antibody type:Primary
- Antigen symbol:CstF-50
- Clonality:Polyclonal
- Clone:R3
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminus of Human CstF-50.
CstF-50 (R3) pAb detects endogenous levels of CstF-50 protein
Type: Primary
Antigen: CstF-50
Clonality: Polyclonal
Clone: R3
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat