Anti-p-ATF2 Rabbit Polyclonal Antibody [clone: T69]
Codice catalogo BWRLBS4017
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-ATF2
- Clonality:Polyclonal
- Clone:T69
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human ATF2 around the phosphorylation site of Threonine 69.
p-ATF2 (T69) pAb detects endogenous levels of ATF2 protein only when phosphorylated at Thr69.
Type: Primary
Antigen: p-ATF2
Clonality: Polyclonal
Clone: T69
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat