Anti-Annexin III Rabbit Polyclonal Antibody [clone: Y64]
Catalog # BWRLBS3519
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Annexin III
- Clonality:Polyclonal
- Clone:Y64
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 41-70 of Human Annexin III.
Annexin III (Y64) pAb detects endogenous levels of Annexin III protein.
Type: Primary
Antigen: Annexin III
Clonality: Polyclonal
Clone: Y64
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat