Order Entry
Italy
ContactUsLinkComponent
Anti-ACTR-IB Rabbit Polyclonal Antibody [clone: G111]
Anti-ACTR-IB Rabbit Polyclonal Antibody [clone: G111]
Catalog # BWRLBS1988
Anti-ACTR-IB Rabbit Polyclonal Antibody [clone: G111]
Catalog # BWRLBS1988

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ACTR-IB
  • Clonality:
    Polyclonal
  • Clone:
    G111
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 100-150 of Human ACTR-IB.

ACTR-IB (G111) pAb detects endogenous levels of ACTR-IB protein.

Type: Primary
Antigen: ACTR-IB
Clonality: Polyclonal
Clone: G111
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat