Order Entry
Italy
ContactUsLinkComponent
Anti-CstF-64 Rabbit Polyclonal Antibody [clone: R46]
Anti-CstF-64 Rabbit Polyclonal Antibody [clone: R46]
Catalog # BWRLBS1980
Anti-CstF-64 Rabbit Polyclonal Antibody [clone: R46]
Catalog # BWRLBS1980

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CstF-64
  • Clonality:
    Polyclonal
  • Clone:
    R46
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids N-terminus of Human CstF-64.

CstF-64 (R46) pAb detects endogenous levels of CstF-64 protein.

Type: Primary
Antigen: CstF-64
Clonality: Polyclonal
Clone: R46
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat