Anti-HLA-H Rabbit Polyclonal Antibody [clone: I104]
Catalog # BWRLBS3876
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:HLA-H
- Clonality:Polyclonal
- Clone:I104
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 72-126 of Human HLA-H.
HLA-H (I104) pAb detects endogenous levels of HLA-H protein.
Type: Primary
Antigen: HLA-H
Clonality: Polyclonal
Clone: I104
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human