Anti-IL-20Ralpha Rabbit Polyclonal Antibody [clone: V222]
Catalog # BWRLBS3472
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IL-20R alpha
- Clonality:Polyclonal
- Clone:V222
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 190-240 of Human IL-20Ralpha.
IL-20R? (V222) pAb detects endogenous levels of IL-20R? protein.
Type: Primary
Antigen: IL-20Ralpha
Clonality: Polyclonal
Clone: V222
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human