Anti-ACCalpha Rabbit Polyclonal Antibody [clone: K86]
Catalog # BWRLBS1377
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ACC alpha
- Clonality:Polyclonal
- Clone:K86
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 50-100 of Human ACCalpha.
ACC? (H74) pAb detects endogenous levels of ACC? protein.
Type: Primary
Antigen: ACCalpha
Clonality: Polyclonal
Clone: K86
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat