Anti-CD11B Rabbit Polyclonal Antibody [clone: E1 ]
BWRLAP0752
- Antibody type:Primary
- Antigen symbol:CD11B
- Clonality:Polyclonal
- Clone:E1
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human CD11B (339aa-629aa).
CD11B (E1) pAb detects endogenous levels of CD11B protein.
Type: Primary
Antigen: CD11B
Clonality: Polyclonal
Clone: E1
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat