Anti-Smad2/3 Rabbit Polyclonal Antibody [clone: N175]
Catalog # BWRLAP0445
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Smad2/3
- Clonality:Polyclonal
- Clone:N175
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 145-195 of Human Smad2.
Smad2/3 (N175) pAb detects endogenous levels of Smad2/3 protein.
Type: Primary
Antigen: Smad2/3
Clonality: Polyclonal
Clone: N175
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat