Anti-E2F4/E2F5 Rabbit Polyclonal Antibody [clone: L50/83]
Catalog # BWRLBS1095
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:E2F4/E2F5
- Clonality:Polyclonal
- Clone:L50/83
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 18-72 of Human E2F4.
E2F4/E2F5 (L50/83) pAb detects endogenous levels of E2F4/E2F5 protein.
Type: Primary
Antigen: E2F4/E2F5
Clonality: Polyclonal
Clone: L50/83
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat